Learn More
Research Products International Corp Sulfamethoxazole, 25 Grams
SDP
Catalog No. 50213673
Encompass_Preferred
Sulfamethoxazole is an antimicrobial agent. It belongs to the class of sulfonamide antibiotics. It acts by inhibiting bacterial folate synthesis, specifically by blocking the enzyme dihydropteroate synthase. This disruption in folate production hinders bacterial growth and replication. 
Sulfamethoxazole may be used to study bacterial susceptibility patterns or as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.
Catalog No. 50-213-673 Supplier Research Products International Corp Supplier No. S4700025.0
Sulfamethoxazole is an antimicrobial agent. It belongs to the class of sulfonamide antibiotics. It acts by inhibiting bacterial folate synthesis, specifically by blocking the enzyme dihydropteroate synthase. This disruption in folate production hinders bacterial growth and replication. 
Sulfamethoxazole may be used to study bacterial susceptibility patterns or as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic.