Learn More
Medchemexpress LLC Sulfamethoxazole-d4 | 1020719-86-1 | 99.3% | 1 MG
SDP
Catalog No. 5000406810
Encompass_Preferred
Sulfamethoxazole-d4 is a deuterium-labeled sulfonamide antibiotic with broad antibacterial activity. It functions by inhibiting bacterial folate metabolism through competition with 4-Aminobenzoic acid (PABA) for dihydropteroate synthetase and dihydropteroate reductase. This compound is suitable for studies of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Available in solid form
Catalog No. 50-004-06810 Supplier Medchemexpress LLC Supplier No. HYB0322S1MG
Sulfamethoxazole-d4 is a deuterium-labeled sulfonamide antibiotic with broad antibacterial activity. It functions by inhibiting bacterial folate metabolism through competition with 4-Aminobenzoic acid (PABA) for dihydropteroate synthetase and dihydropteroate reductase. This compound is suitable for studies of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Available in solid form