Learn More
Medchemexpress LLC Sulfamethoxazole-d4 | 1020719-86-1 | 99.3% | 1 ML
SDP
Catalog No. 5000406345
Encompass_Preferred
Sulfamethoxazole-d4 is a deuterium-labeled sulfonamide antibiotic, also known as Ro 4-2130-d4. It demonstrates widespread antibacterial activity by inhibiting bacterial folate metabolism, specifically by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase. This product is intended for research use only.
  • Deuterium labeled analog
  • Sulfonamide antibiotic with widespread antibacterial activity
  • Inhibits bacterial folate metabolism
  • Suitable for studying urinary tract infections (UTIs), prostatitis, and bronchitis
  • Supplied as a 10 mM solution in DMSO
Catalog No. 50-004-06345 Supplier Medchemexpress LLC Supplier No. HYB0322S10MM/1ML
Sulfamethoxazole-d4 is a deuterium-labeled sulfonamide antibiotic, also known as Ro 4-2130-d4. It demonstrates widespread antibacterial activity by inhibiting bacterial folate metabolism, specifically by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase. This product is intended for research use only.
  • Deuterium labeled analog
  • Sulfonamide antibiotic with widespread antibacterial activity
  • Inhibits bacterial folate metabolism
  • Suitable for studying urinary tract infections (UTIs), prostatitis, and bronchitis
  • Supplied as a 10 mM solution in DMSO