Learn More
Medchemexpress LLC Sulfamethoxazole-d4 | 1020719-86-1 | MFCD03425632 | 98.2% | 257.30 g/mol | C10H7D4N3O3S | 10 MG
SDP
Catalog No. 5000297071
Encompass_Preferred
Sulfamethoxazole-d4 is a deuterium-labeled analogue of the sulfonamide antibiotic sulfamethoxazole, supplied as a high-purity powder for analytical and tracer applications. Intended as an isotope-labeled internal standard for quantitative LC-MS and related studies; molecular formula C10H7D4N3O3S, molecular weight 257.30 g/mol, CAS 1020719-86-1.
  • Deuterium-labeled internal standard for quantitative LC-MS analysis.
  • High purity suitable for LC-MS and other analytical methods.
  • Powder form for easy dissolution and aliquoting.
  • Stable under recommended storage conditions for long-term use.
  • Applicable to studies of bacterial folate metabolism and related assays.
Catalog No. 50-002-97071 Supplier Medchemexpress LLC Supplier No. HYB0322S10MG
Sulfamethoxazole-d4 is a deuterium-labeled analogue of the sulfonamide antibiotic sulfamethoxazole, supplied as a high-purity powder for analytical and tracer applications. Intended as an isotope-labeled internal standard for quantitative LC-MS and related studies; molecular formula C10H7D4N3O3S, molecular weight 257.30 g/mol, CAS 1020719-86-1.
  • Deuterium-labeled internal standard for quantitative LC-MS analysis.
  • High purity suitable for LC-MS and other analytical methods.
  • Powder form for easy dissolution and aliquoting.
  • Stable under recommended storage conditions for long-term use.
  • Applicable to studies of bacterial folate metabolism and related assays.