Learn More
Cayman Chemical Sulfamethoxazole-d4, Purity: ≥99% deuterated forms (d1-d4); ≤1% d0, CAS: 1020719-86-1, 1 mg
SDP
Catalog No. 502941962
Encompass_Preferred
An internal standard for the quantification of sulfamethoxazole by GC- or LC-MS
Catalog No. 50-294-1962 Supplier Cayman Chemical Supplier No. 286181 MG
An internal standard for the quantification of sulfamethoxazole by GC- or LC-MS