Learn More
Cayman Chemical Sulfamethoxazole-d4 5mg
SDP
Catalog No. 502938242
Encompass_Preferred
An internal standard for the quantification of sulfamethoxazole by GC- or LC-MS
Catalog No. 50-293-8242 Supplier Cayman Chemical Supplier No. 286185 MG
An internal standard for the quantification of sulfamethoxazole by GC- or LC-MS