Learn More
AdipoGen Sulfamethoxazole hydroxylamine
SDP
Catalog No. 502560072
Encompass_Preferred
Chemical. CAS 114438-33-4. Formula C10H11N3O4S. MW 269.28. Synthetic Metabolite of sulfamethoxazole, a sulfonamide bacteriostatic antibiotic. Has been shown to be an important component in sulfonamide hypersensitivity reactions and has been implicated in sulfonamide-mediated immunomodulatory effects. Shown to induce lymphocyte toxicity and suppression of proliferation. Compound can be used as analytical reference material.
Catalog No. 50-256-0072 Supplier AdipoGen Supplier No. CDXS0056M010
Chemical. CAS 114438-33-4. Formula C10H11N3O4S. MW 269.28. Synthetic Metabolite of sulfamethoxazole, a sulfonamide bacteriostatic antibiotic. Has been shown to be an important component in sulfonamide hypersensitivity reactions and has been implicated in sulfonamide-mediated immunomodulatory effects. Shown to induce lymphocyte toxicity and suppression of proliferation. Compound can be used as analytical reference material.