Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethoxazole, MilliporeSigma™ Supelco™

Catalog No. 111015357 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5357 100 g
11-101-5356 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5357 Supplier MilliporeSigma Supelco Supplier No. S7507100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfamethoxazole is a bacteriostatic antibiotic, belonging to the class of sulfonamides. It is widely used in the treatment of pneumocystis, coccidiosis, gastroenteritis, diarrhea, respiratory diseases such as pneumonia, etc.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 723-46-6
Physical Form Neat
Linear Formula C10H11N3O3 S
MDL Number MFCD00010546
Synonym 4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide
Type Analytical Standard
Quantity 100 g
Formula Weight 253.28
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.