Learn More
Medchemexpress LLC Sulfamethoxazole (Ro 4-2130) | 723-46-6 | 99.7% | 250 MG
SDP
Catalog No. 5000405732
Encompass_Preferred
Sulfamethoxazole is a sulfonamide antibiotic with widespread antibacterial activity. It inhibits bacterial folate metabolism by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase. It can be used for the study of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Widespread antibacterial activity
  • Inhibits bacterial folate metabolism
  • Used for the study of urinary tract infections, prostatitis, and bronchitis
  • Inhibits P. lutzii mitochondrial activity
  • Inhibits KATP channel currents in mouse pancreatic β cells
  • Stimulates insulin secretion in mouse islets
Catalog No. 50-004-05732 Supplier Medchemexpress LLC Supplier No. 250MGHYB0322R
Add to Cart
Add to Cart
Sulfamethoxazole is a sulfonamide antibiotic with widespread antibacterial activity. It inhibits bacterial folate metabolism by competing with 4-Aminobenzoic acid (PABA) to act on dihydropteroate synthetase and dihydropteroate reductase. It can be used for the study of urinary tract infections (UTIs), prostatitis, and bronchitis.
  • Widespread antibacterial activity
  • Inhibits bacterial folate metabolism
  • Used for the study of urinary tract infections, prostatitis, and bronchitis
  • Inhibits P. lutzii mitochondrial activity
  • Inhibits KATP channel currents in mouse pancreatic β cells
  • Stimulates insulin secretion in mouse islets