Learn More
Cayman Chemical Sulfamethoxypyridazine, Purity: ≥95%, CAS: 80-35-3, 10 g
SDP
Catalog No. 502986033
Encompass_Preferred
A sulfonamide antibiotic that inhibits the growth of Gram-negative bacteria and other microorganisms; inhibits recombinant P. carinii DHPS (IC50 = 17 nM); inhibits folate biosynthesis in P. carinii by 75% at 50 nM; inhibits any visible growth in culture tubes of 378 out of 530 E. coli isolates from necropsied animals and poultry at 0.25 mg/ml; decreases the number of viable amoxicillin-induced B. burgdorferi round body forms to 30% of control at 50 μM; decreases the number of P. carinii trophozoite and cyst lesions in lung tissue in a mouse model of pneumonia infection (ED50s = 0.06 and 0.08 mg/kg per day, respectively)
Catalog No. 50-298-6033 Supplier Cayman Chemical Supplier No. 2187510 G
A sulfonamide antibiotic that inhibits the growth of Gram-negative bacteria and other microorganisms; inhibits recombinant P. carinii DHPS (IC50 = 17 nM); inhibits folate biosynthesis in P. carinii by 75% at 50 nM; inhibits any visible growth in culture tubes of 378 out of 530 E. coli isolates from necropsied animals and poultry at 0.25 mg/ml; decreases the number of viable amoxicillin-induced B. burgdorferi round body forms to 30% of control at 50 μM; decreases the number of P. carinii trophozoite and cyst lesions in lung tissue in a mouse model of pneumonia infection (ED50s = 0.06 and 0.08 mg/kg per day, respectively)