Learn More
Medchemexpress LLC Sulfamethoxypyridazine | 80-35-3 | 99.9% | 250 MG
SDP
Catalog No. 5000414356
Encompass_Preferred
Sulfamethoxypyridazine is a long-acting sulfonamide antibiotic primarily used for the treatment of Dermatitis herpetiformis. It is intended for research use only and not for sale to patients. This compound is available in solid form and is stable under recommended storage conditions, making it suitable for various laboratory applications.
  • Used for treatment of dermatitis herpetiformis
  • Long-acting sulfonamide antibiotic
  • Intended for research use only
  • Stable under recommended storage conditions
Catalog No. 50-004-14356 Supplier Medchemexpress LLC Supplier No. 250MGHYB1387
Add to Cart
Add to Cart
Sulfamethoxypyridazine is a long-acting sulfonamide antibiotic primarily used for the treatment of Dermatitis herpetiformis. It is intended for research use only and not for sale to patients. This compound is available in solid form and is stable under recommended storage conditions, making it suitable for various laboratory applications.
  • Used for treatment of dermatitis herpetiformis
  • Long-acting sulfonamide antibiotic
  • Intended for research use only
  • Stable under recommended storage conditions