Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sulfamic Acid
SCI_ED

Catalog No. S25792 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25792 50 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25792
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Density 2.15 g/mL
Packaging Solid
Quantity 50 g
Linear Formula H2NSO3H
Formula Weight 97.09
Chemical Name or Material Sulfamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.