Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic acid, ≥99.5% (alkalimetric), Solstice

Catalog No. 60044601 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
6 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-601 6 x 2.5 kg
60-015-66 1 kg
60-015-67 2.5 kg
60-015-68 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-601 Supplier Solstice Supplier No. 074016X2.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

≥99.5% (alkalimetric)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Residue after Ignition ≤0.02%
Quantity 6 x 2.5 kg
Linear Formula NH2SO3H
UN Number UN2967
Formula Weight 97.09
Percent Purity ≥99.5% (alkalimetric)
Chemical Name or Material Sulfamic Acid
Hazard Statement P273-P305 + P351 + P338
Precautionary Statement H315-H319-H412
DOTInformation 8 / PGIII
EINECSNumber 226-218-8
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.