Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, 99+%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19486080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19486080 100 g
ICN19486090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19486080 Supplier MP Biomedicals, Inc Supplier No. 0219486080
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Assay Percent Range 99+%
Quantity 100 g
Formula Weight 97.09
Grade ACS Reagent
Chemical Name or Material Sulfamic Acid
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.