Learn More
Sigma Aldrich Fine Chemicals Biosciences Sulfamic acid ACS reagent500G
Catalog No. 502891481
Encompass
Sulfamic acid is a strong inorganic acid. It is a hypochlorite scavenger. The ability of sulfamic acid to reduce nitrate to nitrogen gas under acidic condition has been utilized to denitrify nitrate-rich wastewater along with zinc powder.
Catalog No. 50-289-1481 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 383120500G
Sulfamic acid is a strong inorganic acid. It is a hypochlorite scavenger. The ability of sulfamic acid to reduce nitrate to nitrogen gas under acidic condition has been utilized to denitrify nitrate-rich wastewater along with zinc powder.