Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS110025A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS110025A 2.5 kg Natural Poly Bottle
RDCS1100100 100 g White Poly Wide Mouth Bottle
RDCS1100500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RDCS110025A Supplier Ricca Chemical Company Supplier No. RDCS11002.5A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging Natural Poly Bottle
Residue after Ignition max. 0.01%
Quantity 2.5 kg
Physical Form Crystals
Synonym amidosulfonic acid
SMILES NS(O)(=O)=O
Molecular Weight (g/mol) 97.09
Percent Purity 99.3 to 100.3%
Grade ACS
Chemical Name or Material Sulfamic Acid
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.