Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid (Crystalline/Certified ACS), Fisher Chemical

Catalog No. A295500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A295-500 500 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A295-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

pH 1.2
Color White
Melting Point 200°C
Density 2.15 g/cm3
Identification Pass Test
Packaging Amber Glass
Assay Percent Range 99.3 to 100.3 %
Quantity 500 g
Physical Form Solid
Linear Formula NH2SO3H
Solubility Information 0.010% max. Insoluble Matter
Formula Weight 97.09
Percent Purity 99.3 to 100.3%
Grade Certified ACS
Chemical Name or Material Sulfamic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Corrosive to metals. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.