Learn More
Hach Company Sulfamic Acid Powder Pillows, pk/100
Supplier: Hach Company 107399
For determination of Dissolved Oxygen by the Winkler titration method. Range: 1 to 10 mg/L. Hach method 8229. Sample size: 300 mL.