Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamonomethoxine, 98%

Catalog No. AAJ6673303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6673303 1 g
AAJ66733MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6673303 Supplier Thermo Scientific Chemicals Supplier No. J6673303
Add to Cart

Long-acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica. | CAS: 1220-83-3 | C11H12N4O3S | 280.302 g/mol

Sulfamonomethoxine is a long acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1220-83-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.302
MDL Number MFCD00063466
InChI Key WMPXPUYPYQKQCX-UHFFFAOYSA-N
Synonym sulfamonomethoxine, sulfamonomethoxin, daimeton, sulfamonometoxina, sulfamonomethoxinum, 4-amino-n-6-methoxypyrimidin-4-yl benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl benzenesulfonamide, benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl, n1-6-methoxy-4-pyrimidinyl sulfanilamide, sulfamonomethoxinum inn-latin
PubChem CID 5332
ChEBI CHEBI:32164
IUPAC Name 4-amino-N-(6-methoxypyrimidin-4-yl)benzenesulfonamide
SMILES COC1=NC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 203°C to 206°C
Boiling Point 513°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 1 g
Beilstein 621128
Solubility Information Soluble in water at 10mg/ml with warming. Also soluble in ethanol or methanol
Formula Weight 280.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulfamonomethoxine
Show More Show Less
Hazard Category H315-H317-H319-H335
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
RTECSNumber WP0550000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.