Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamonomethoxine, G-Biosciences
SCI_ED

Catalog No. S139828
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139828 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139828 Supplier G-Biosciences Supplier No. RC1108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfamonomethoxine is a long-acting sulfonamide antibiotic.

It is used in blood kinetic studies as well as to study the formation of capsules in Bordetella bronchiseptica. Sulfamonomethoxine is used to combat hyperpyrexia of unknown etiology.

Chemical Identifiers

Molecular Weight (g/mol) 280.3

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Boiling Point 513.2°C
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.