Learn More
ALADDIN SCIENTIFIC CORPORATION Sulfanilamide, 100g
Catalog No. 5000082506
Encompass_Preferred
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid It serves as an intermediate for the preparation of 26-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid
Catalog No. 50-000-82506 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. S108473100G
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid It serves as an intermediate for the preparation of 26-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid