Learn More
Medchemexpress LLC Sulfanilamide | 63-74-1 | 99.3% | 1 G
SDP
Catalog No. 5000405862
Encompass_Preferred
Sulfanilamide is a potent and orally active sulfonamide antibiotic, also serving as a major intermediate in sulfamethoxazole biodegradation. This compound acts as a carbonic anhydrase inhibitor and has demonstrated inhibition against the virus of lymphogranuloma venereum. It presents as a white to off-white solid and is intended solely for research and analytical applications.
  • Potent and orally active sulfonamide antibiotic properties
  • Functions as a carbonic anhydrase inhibitor
  • Exhibits inhibition against the virus of lymphogranuloma venereum
  • Store powder at -20°C for up to 3 years, or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 2 years, or -20°C for up to 1 year
Catalog No. 50-004-05862 Supplier Medchemexpress LLC Supplier No. HYB02421G
Sulfanilamide is a potent and orally active sulfonamide antibiotic, also serving as a major intermediate in sulfamethoxazole biodegradation. This compound acts as a carbonic anhydrase inhibitor and has demonstrated inhibition against the virus of lymphogranuloma venereum. It presents as a white to off-white solid and is intended solely for research and analytical applications.
  • Potent and orally active sulfonamide antibiotic properties
  • Functions as a carbonic anhydrase inhibitor
  • Exhibits inhibition against the virus of lymphogranuloma venereum
  • Store powder at -20°C for up to 3 years, or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 2 years, or -20°C for up to 1 year