Learn More
Medchemexpress LLC Sulfanilamide | 63-74-1 | MFCD00007939 | 99.9% | 10 MM IN 1 ML
SDP
Catalog No. 5000406956
Encompass_Preferred
Sulfanilamide is a potent and orally active sulfonamide antibiotic. It is a major intermediate of sulfamethoxazole biodegradation. It also functions as a carbonic anhydrase inhibitor and exhibits inhibition against the virus of lymphogranuloma venereum.
  • Potent anti-infection activity in albino mice
  • Inhibits HepG2 cells with an IC50 of 0.08 μM
  • Inhibits K562 cells with an IC50 of 0.2 μM
  • Inhibits MDA-MB-231 cells with an IC50 of 0.09 μM
Catalog No. 50-004-06956 Supplier Medchemexpress LLC Supplier No. HYB024210MM/1ML
Sulfanilamide is a potent and orally active sulfonamide antibiotic. It is a major intermediate of sulfamethoxazole biodegradation. It also functions as a carbonic anhydrase inhibitor and exhibits inhibition against the virus of lymphogranuloma venereum.
  • Potent anti-infection activity in albino mice
  • Inhibits HepG2 cells with an IC50 of 0.08 μM
  • Inhibits K562 cells with an IC50 of 0.2 μM
  • Inhibits MDA-MB-231 cells with an IC50 of 0.09 μM