Learn More
Sigma Aldrich Fine Chemicals Biosciences Sulfanilamide, 63-74-1, MFCD00007939, 500 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S9251500G
Encompass
Linear Formula: H2NC6H4SO2NH2, Molecular Weight: 172.20, ≥99%, Synonym: p-Aminobenzenesulfonamide. Sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase.
Catalog No. NC1070272