Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilamide, MP Biomedicals™

Catalog No. MP210298880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210298880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210298880 Supplier MP Biomedicals, Inc Supplier No. 0210298880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as antibacterial agent and to treat vaginal fungal infections. Sulfanilamide, MP Biomedicals is a sulfonamide antibiotic that blocks the synthesis of folic acid by inhibiting the enzyme dihydropteroate synthase. Effective against gram-negative bacter

Sulfanilamide is a molecule containing the sulfonamide functional group attached to an aniline. As a sulfonamide antibiotic, it functions by competitively inhibiting enzymatic reactions (i.e., acting as a substrate analogue) involving para-aminobenzoic acid (PABA). PABA is needed in enzymatic reactions that produce folic acid, which acts as a coenzyme in the synthesis of amino acids. By inhibiting dihydropteroate synthase, it blocks the synthesis of folic acid.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Absorbance λ max (water): 255–259nm
Melting Point 160°C to 170°C
Density 1.08g/mL (Lit.)
Quantity 100 g
Solubility Information Soluble in 1g dissolves in 37mL alcohol, 5mL acetone; glycerol, ether, benzene, petroleum ether.
Formula Weight 172.2
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.