Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilamide/N-(Naphthyl) Reagent, For Nitrogen (Nitrate/Nitrite), LabChem™

Catalog No. LC254602 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC254602 1 L
LC254601 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC254602 Supplier LabChem, Inc. Supplier No. LC254602
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Sulfanilamide/N-(Naphthyl) Reagent
Color Pink to Tan
Name Note For Nitrogen (Nitrate/Nitrite)
CAS Max % 85.39, 13.51, 1, 0.1
CAS 7664-38-2, 63-74-1, 1465-25-4
Linear Formula C10H7NHCH2CH2NH2·2HCl
Density 1.07g/mL
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 259.18
Identification Passes Test
Packaging Amber Poly
Specific Gravity 1.07g/mL
Viscosity 1.35cSt
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Phosphorus oxides
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.