Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilamide Reagent, 1% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
SDP

Catalog No. 810332 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8103-32 1 L Natural Poly Bottle
8103-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8103-32 Supplier Ricca Chemical Company Supplier No. 810332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63-74-1 , 7647-01-0 , 7732-18-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Color Colorless
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix 10% (v/v) Hydrochloric Acid
pH <1
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Sulfanilamide Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.