Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Sulfanilic Acid

Catalog No. 01549970 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-970 30 mL
01-549-997 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-549-970 Supplier Beckman Coulter™ Supplier No. B101044A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sulfanilic Acid for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.8%
For Use With (Equipment) Beckman Coulter Analyzers
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.