Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic Acid, Anhydrous, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18614171 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-171 50 kg Poly Bottle
18-614-170 500 g Poly Bottle
18-614-169 12 kg Poly Bottle
18-614-172 25 g Poly Bottle
18-614-168 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-171 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S170350KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
IUPAC Name 4-aminobenzene-1-sulfonic acid
SMILES NC1=CC=C(C=C1)S(O)(=O)=O

Specifications

CAS Max % 1
Boiling Point 300°C
Ignition Residue 0.0001
Packaging Poly Bottle
Quantity 50 kg
Percent Purity 98 to 102%
Grade ACS
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.