Learn More
FUJIFILM Biosciences Sulfanilic Acid Azo GSalt STND
Catalog No. 5000450393
Encompass_Preferred
High-purity sulfanilic acid azo compound used as a color standard in food additive testing red to reddish yellow powder It is the impurities that may be included in the Food Yellow No 5 Y5 and Food Yellow No 5 Aluminum Lake Y5Al Azo dye-based reference standard for determining color properties in analytical procedures for food additives meets purity and spectral criteria
Catalog No. 50-004-50393 Supplier FUJIFILM Biosciences Supplier No. 19817501
High-purity sulfanilic acid azo compound used as a color standard in food additive testing red to reddish yellow powder It is the impurities that may be included in the Food Yellow No 5 Y5 and Food Yellow No 5 Aluminum Lake Y5Al Azo dye-based reference standard for determining color properties in analytical procedures for food additives meets purity and spectral criteria