Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012775 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2775 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2775 Supplier MilliporeSigma Supelco Supplier No. PHR1419500MG
Add to Cart
Add to Cart

Sulfanilic Acid is a synthetic organic compound, obtained via sulfonation of aniline with sulfuric acid. It is widely used for the production of optical brightening agents, synthetic organic dyes, food colorants and additives.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 121-57-3
Linear Formula 4-(H2 N)C6H4 SO3 H
MDL Number MFCD00007886
Melting Point >300°C
Synonym Sulfanilic acid; 4-Aminobenzenesulfonic acid; Aniline-4-sulfonic acid
Type Certified Reference Material
Quantity 500 mg
Formula Weight 173.19
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.