Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfaphenazole | 526-08-9 | 99.8% | 314.36 g/mol | C15H14N4O2S | 10 MG
SDP

Catalog No. 5000298203
Encompass_Preferred

Sulfaphenazole is an analytical reference standard intended for research and analytical applications; it is a selective inhibitor of CYP2C9 and supplied in calibrated milligram quantities with batch documentation.

  • Analytical standard for assay calibration and reference
  • Selective CYP2C9 inhibitor used in metabolism studies
  • High stated purity (99.8%) for accurate quantitation
  • Molecular weight 314.36 g/mol; formula C15H14N4O2S
  • Supplied with certificate of analysis and safety data sheet for batch-specific information

Catalog No. 50-002-98203 Supplier Medchemexpress LLC Supplier No. HYB1218R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfaphenazole is an analytical reference standard intended for research and analytical applications; it is a selective inhibitor of CYP2C9 and supplied in calibrated milligram quantities with batch documentation.

  • Analytical standard for assay calibration and reference
  • Selective CYP2C9 inhibitor used in metabolism studies
  • High stated purity (99.8%) for accurate quantitation
  • Molecular weight 314.36 g/mol; formula C15H14N4O2S
  • Supplied with certificate of analysis and safety data sheet for batch-specific information

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.