Learn More
Medchemexpress LLC Sulfapyridine | 144-83-2 | 99.85% | 10 G
SDP
Catalog No. 5000405642
Encompass_Preferred
Sulfapyridine is a sulfonamide antibiotic agent and a major metabolite of Sulfasalazine. It inhibits recombinant *P. carinii* dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. This compound also exhibits antibacterial, anti-inflammatory, and anti-rheumatic activities.
  • Sulfonamide antibiotic agent
  • Major metabolite of Sulfasalazine
  • Inhibits recombinant *P. carinii* dihydropteroate synthetase (DHPS)
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-rheumatic activities
Catalog No. 50-004-05642 Supplier Medchemexpress LLC Supplier No. HYB021210G
Sulfapyridine is a sulfonamide antibiotic agent and a major metabolite of Sulfasalazine. It inhibits recombinant *P. carinii* dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. This compound also exhibits antibacterial, anti-inflammatory, and anti-rheumatic activities.
  • Sulfonamide antibiotic agent
  • Major metabolite of Sulfasalazine
  • Inhibits recombinant *P. carinii* dihydropteroate synthetase (DHPS)
  • Exhibits antibacterial activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-rheumatic activities