Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfapyridine | 144-83-2 | 99.9% | 5 G
SDP

Supplier:  Medchemexpress LLC HYB02125G

Encompass_Preferred

Sulfapyridine is a sulfonamide antibiotic agent and a significant metabolite of Sulfasalazine. It exhibits antibacterial, anti-inflammatory, and anti-rheumatic activities by inhibiting recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM.

  • Inhibits production of IL-8, CXCL1, and MCP-1 in synovial cells.
  • Effective against Y. enterocolitica and Salmonella.
  • Significantly inhibits systemic allergic reactions in rats.
  • Molecular weight: 249.29.
  • Chemical formula: C11H11N3O2S.
  • Appearance: white to off-white solid.

Catalog No. 50-004-06400


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.