Learn More
Medchemexpress LLC Sulfapyridine | 144-83-2 | MFCD00038036 | 99.9% | 1 G
SDP
Catalog No. 5000406349
Encompass_Preferred
Sulfapyridine is a sulfonamide antibiotic that is a major metabolite of Sulfasalazine. It acts as an antibiotic agent by inhibiting recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. Beyond its antibacterial properties, Sulfapyridine also exhibits anti-inflammatory and anti-rheumatic activities.
  • Inhibits IL-8, CXCL1, and MCP-1 production in synovial cells
  • Active against Y. enterocolitica and Salmonella
  • Significantly inhibits systemic allergic reactions in rats
  • For research use only
Catalog No. 50-004-06349 Supplier Medchemexpress LLC Supplier No. HYB02121G
Sulfapyridine is a sulfonamide antibiotic that is a major metabolite of Sulfasalazine. It acts as an antibiotic agent by inhibiting recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. Beyond its antibacterial properties, Sulfapyridine also exhibits anti-inflammatory and anti-rheumatic activities.
  • Inhibits IL-8, CXCL1, and MCP-1 production in synovial cells
  • Active against Y. enterocolitica and Salmonella
  • Significantly inhibits systemic allergic reactions in rats
  • For research use only