Learn More
Medchemexpress LLC HY-B0212 500mg Medchemexpress, Sulfapyridine CAS:144-83-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0212500MG
Encompass_Preferred
Medchemexpress, HY-B0212 500mg Sulfapyridine CAS:144-83-2 Sulfapyridine, a major metabolite of Sulfasalazine, is a sulfonamide antibiotic agent. Sulfapyridine inhibits recombinant P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM. Sulfapyridine has antibacterial, anti-inflammatory and anti-rheumatic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2411
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.