Learn More
Apexbio Technology LLC Sulfapyridine 144-83-2 5g
SDP
Catalog No. 502240420
Encompass_Preferred
Sulfapyridine (144-83-2) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It functions by inhibiting this enzyme thus blocking folic acid synthesis and suppressing bacterial growth Sulfapyridine exerts its biological activity primarily through inhibition of bacterial dihydropteroate synthase In in vitro studies sulfapyridine demonstrates inhibitory activity with IC50 values typically ranging from approximately 5 g/mL to 20 g/mL depending on the bacterial strain and assay conditions Based on these pharmacological properties sulfapyridine holds research potential in antimicrobial susceptibility testing investigations of bacterial resistance mechanisms and studies of bacterial folate pathway enzymes
This item is not returnable. View return policy
Catalog No. 50-224-0420 Supplier Apexbio Technology LLC Supplier No. B18445000
Sulfapyridine (144-83-2) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It functions by inhibiting this enzyme thus blocking folic acid synthesis and suppressing bacterial growth Sulfapyridine exerts its biological activity primarily through inhibition of bacterial dihydropteroate synthase In in vitro studies sulfapyridine demonstrates inhibitory activity with IC50 values typically ranging from approximately 5 g/mL to 20 g/mL depending on the bacterial strain and assay conditions Based on these pharmacological properties sulfapyridine holds research potential in antimicrobial susceptibility testing investigations of bacterial resistance mechanisms and studies of bacterial folate pathway enzymes