Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfapyridine, 95%, Thermo Scientific Chemicals

Catalog No. AC442850250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442850250 25 g
AC442851000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC442850250 Supplier Thermo Scientific Chemicals Supplier No. 442850250
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-83-2
Molecular Formula C11H11N3O2S
Molecular Weight (g/mol) 249.288
InChI Key GECHUMIMRBOMGK-UHFFFAOYSA-N
PubChem CID 5336
ChEBI CHEBI:132842
IUPAC Name 4-amino-N-pyridin-2-ylbenzenesulfonamide
SMILES C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Type Sulfapyridine
Melting Point 190.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Quantity 25 g
Formula Weight 249.29
Percent Purity 95%
Chemical Name or Material Sulfapyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.