Learn More
Medchemexpress LLC Sulfapyridine (Standard) | 144-83-2 | 99.9% | 100 MG
SDP
Catalog No. 5000405746
Encompass_Preferred
Sulfapyridine (Standard) is an analytical standard intended for research and analytical applications. This compound is a major metabolite of Sulfasalazine and functions as a sulfonamide antibiotic agent, exhibiting antibacterial, anti-inflammatory, and anti-rheumatic activities. It serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Functions as a sulfonamide antibiotic agent
  • Exhibits antibacterial, anti-inflammatory, and anti-rheumatic activities
  • Inhibits P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM
  • Used in HPLC, GC, and MS research experiments
  • Inhibits production of IL-8, CXCL1, and MCP-1 in synovial cells of rheumatoid arthritis
  • Inhibits systemic allergic reactions in vivo
Catalog No. 50-004-05746 Supplier Medchemexpress LLC Supplier No. HYB0212R100MG
Sulfapyridine (Standard) is an analytical standard intended for research and analytical applications. This compound is a major metabolite of Sulfasalazine and functions as a sulfonamide antibiotic agent, exhibiting antibacterial, anti-inflammatory, and anti-rheumatic activities. It serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Functions as a sulfonamide antibiotic agent
  • Exhibits antibacterial, anti-inflammatory, and anti-rheumatic activities
  • Inhibits P. carinii dihydropteroate synthetase (DHPS) with an IC50 of 0.18 μM
  • Used in HPLC, GC, and MS research experiments
  • Inhibits production of IL-8, CXCL1, and MCP-1 in synovial cells of rheumatoid arthritis
  • Inhibits systemic allergic reactions in vivo