Learn More
Apexbio Technology LLC SULFAQUINOXALINE-SODIUM-200MG
SDP
Catalog No. 5000569511
Encompass_Preferred
Sulfaquinoxaline sodium (CAS No 967-80-6) is a sulfonamide antimicrobial agent primarily used in veterinary medicine research Originally derived from sulfonamide antibiotics it works by inhibiting the folic acid synthesis pathway in bacteria specifically targeting dihydropteroate synthase This action impedes bacterial cell growth and replication In vitro it demonstrates activity in the nanomolar to low micromolar range Typical research applications include evaluating its efficacy in various cell models and animal studies to understand its pharmacokinetics and dynamics It s also used in drug screening processes to assess potential interactions and efficacy in combination therapies Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69511 Supplier Apexbio Technology LLC Supplier No. C7006200
Sulfaquinoxaline sodium (CAS No 967-80-6) is a sulfonamide antimicrobial agent primarily used in veterinary medicine research Originally derived from sulfonamide antibiotics it works by inhibiting the folic acid synthesis pathway in bacteria specifically targeting dihydropteroate synthase This action impedes bacterial cell growth and replication In vitro it demonstrates activity in the nanomolar to low micromolar range Typical research applications include evaluating its efficacy in various cell models and animal studies to understand its pharmacokinetics and dynamics It s also used in drug screening processes to assess potential interactions and efficacy in combination therapies Experimental concentrations typically depend on the specific research design