Learn More
Selleck Chemical LLC Sulfasalazine 10mM/1mL 599-79-1 Azulfidine, Salazopyrin, Sulphasalazine
SDP
Catalog No. 501361401
Encompass_Preferred
Sulfasalazine (Azulfidine, Salazopyrin, Sulphasalazine) is a sulfa derivative of mesalazine, used as an anti-inflammatory agent to treat bowel disease and rheumatoid arthritis. Sulfasalazine is a potent and specific inhibitor of nuclear factor kappa B (NF-?B), TGF-? and COX-2. Sulfasalazine induces ferroptosis, apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1401 Supplier Selleck Chemical LLC Supplier No. S157610MM/1ML
Sulfasalazine (Azulfidine, Salazopyrin, Sulphasalazine) is a sulfa derivative of mesalazine, used as an anti-inflammatory agent to treat bowel disease and rheumatoid arthritis. Sulfasalazine is a potent and specific inhibitor of nuclear factor kappa B (NF-?B), TGF-? and COX-2. Sulfasalazine induces ferroptosis, apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.