Learn More
Medchemexpress LLC HY-14655 500mg , Sulfasalazine CAS:599-79-1 Purity:>98%
SDP
Catalog No. 502260137
Encompass_Preferred
HY-14655 500mg Sulfasalazine CAS: 599-79-1 Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0137 Supplier Medchemexpress LLC Supplier No. HY14655500MG
HY-14655 500mg Sulfasalazine CAS: 599-79-1 Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.