Learn More
Medchemexpress LLC Sulfasalazine | 599-79-1 | 99.6% | 398.39 | 1 ML
SDP
Catalog No. 5000360716
Encompass_Preferred
Sulfasalazine is an anti-rheumatic agent used for the research of rheumatoid arthritis and ulcerative colitis. It functions by suppressing NF-κB activity and is recognized as a type 1 ferroptosis inducer.
  • Suppresses NF-κB activity
  • Induces type 1 ferroptosis
  • Inhibits TNFα-, LPS-, or phorbol ester-induced NFκB activation
  • Prevents TNFα-induced nuclear translocation of NFκB
  • Suppresses glioma growth in vivo
Catalog No. 50-003-60716 Supplier Medchemexpress LLC Supplier No. HY1465510MM/1ML
Sulfasalazine is an anti-rheumatic agent used for the research of rheumatoid arthritis and ulcerative colitis. It functions by suppressing NF-κB activity and is recognized as a type 1 ferroptosis inducer.
  • Suppresses NF-κB activity
  • Induces type 1 ferroptosis
  • Inhibits TNFα-, LPS-, or phorbol ester-induced NFκB activation
  • Prevents TNFα-induced nuclear translocation of NFκB
  • Suppresses glioma growth in vivo