Learn More
Medchemexpress LLC Sulfasalazine | 599-79-1 | >99.6% | 398.39 | 10 G
SDP
Catalog No. 5000360718
Encompass_Preferred
Sulfasalazine is an anti-rheumatic agent investigated for conditions like rheumatoid arthritis and ulcerative colitis. Its mechanism involves suppressing NF-κB activity and acting as a type 1 ferroptosis inducer.
  • Anti-rheumatic properties.
  • Suppresses NF-κB activity.
  • Induces type 1 ferroptosis.
  • Under investigation for primary brain tumors and other conditions.
Catalog No. 50-003-60718 Supplier Medchemexpress LLC Supplier No. HY1465510G
Sulfasalazine is an anti-rheumatic agent investigated for conditions like rheumatoid arthritis and ulcerative colitis. Its mechanism involves suppressing NF-κB activity and acting as a type 1 ferroptosis inducer.
  • Anti-rheumatic properties.
  • Suppresses NF-κB activity.
  • Induces type 1 ferroptosis.
  • Under investigation for primary brain tumors and other conditions.