Learn More
Medchemexpress LLC Sulfasalazine-d4 | 1346606-50-5 | 99.3% | 402.42 | 1 MG
SDP
Catalog No. 5000362944
Encompass_Preferred
Sulfasalazine-d4 is a deuterium-labeled version of Sulfasalazine, an anti-rheumatic agent used in the research of rheumatoid arthritis and ulcerative colitis. It is known to suppress NF-κB activity and functions as a type 1 ferroptosis inducer.
  • Incorporates stable heavy isotopes as tracers
  • Suitable for use as a tracer in various research applications
  • Can be used as an internal standard for quantitative analytical techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-62944 Supplier Medchemexpress LLC Supplier No. HY14655S1MG
Sulfasalazine-d4 is a deuterium-labeled version of Sulfasalazine, an anti-rheumatic agent used in the research of rheumatoid arthritis and ulcerative colitis. It is known to suppress NF-κB activity and functions as a type 1 ferroptosis inducer.
  • Incorporates stable heavy isotopes as tracers
  • Suitable for use as a tracer in various research applications
  • Can be used as an internal standard for quantitative analytical techniques such as NMR, GC-MS, or LC-MS