Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfasalazine, USP, 97-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18607132 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-132 100 g Amber Glass Bottle
18-607-134 500 g Amber Glass Bottle
18-607-133 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-132 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1668100GM
Add to Cart
Add to Cart

S1668, 599-79-1, C18H14N4O5S

Spectrum™ Chemical Sulfasalazine, USP belongs to a class of drugs called sulfa drugs and is used in the treatment of rheumatoid arthritis and some other autoimmune conditions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 599-79-1
Molecular Formula C18H14N4O5S
Molecular Weight (g/mol) 398.39
InChI Key OQANPHBRHBJGNZ-BKUYFWCQSA-N
IUPAC Name (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
SMILES OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

Specifications

CAS Max % 1
Ignition Residue 0.005
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 97 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.