Learn More
INDUSTRIAL TEST SYSTEMS INC SULFATE CHECK TEST 100 USD MIN
SDP
Catalog No. NC3234144
Encompass_Preferred
  • Rapid, dip-and-read method for sulfate detection
  • 30 test strips for convenience and efficiency
  • Ideal for field use and scientific applications
  • Supports monitoring drinking water, industrial wastewater, and other environmental samples
  • Complies with USEPA secondary drinking water standards
  • Maximum sulfate contaminant level of 250 mg/L (ppm)
  • User-friendly design eliminates the need for laboratory equipment
  • Immediate results for on-site analysis
  • Strips decolorize in presence of sulfate for clear visual indication
  • Highly portable, foil-packed strips for easy transportation
Catalog No. NC3234144 Supplier INDUSTRIAL TEST SYSTEMS INC Supplier No. 481200
Add to Cart
Edge
Add to Cart
  • Rapid, dip-and-read method for sulfate detection
  • 30 test strips for convenience and efficiency
  • Ideal for field use and scientific applications
  • Supports monitoring drinking water, industrial wastewater, and other environmental samples
  • Complies with USEPA secondary drinking water standards
  • Maximum sulfate contaminant level of 250 mg/L (ppm)
  • User-friendly design eliminates the need for laboratory equipment
  • Immediate results for on-site analysis
  • Strips decolorize in presence of sulfate for clear visual indication
  • Highly portable, foil-packed strips for easy transportation