Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Sulfate Reducing Bacteria (SRB) Tube Test

Catalog No. 15398343 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-398-343 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-398-343 Supplier Lovibond™ Supplier No. 56B010810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Sulfate Reducing Bacteria (SRB) Tube Test is designed to assess the contamination of water samples with sulfate reducing bacteria. The test contains a straw colored medium which reacts to the production of hydrogen sulfide to give a semi-quantitative result after 5 day.

  • The Sulfate Reducing Bacteria (SRB) test is designed to assess the contamination of water samples with sulfate reducing bacteria.
  • The test contains a straw colored medium which reacts to the production of hydrogen sulfide to give a semi-quantitative result after 5 days.
  • The SRB test is used specifically to indicate the presence of bacteria, which under the correct conditions, are able to produce hydrogen sulfide.
  • Hydrogen sulfide is a colorless gas which is extremely corrosive to ferrous and no ferrous metals.
  • This can lead to holes in water systems and leaks in tankers by dissolving the surrounding metal.

Specifications

Color Straw-colored
Detects Sulfate-reducing Bacteria
Manufacturing Origin United Kingdom
Media Type Semi-solid Medium for The Analysis of Anaerobic Microorganisms That Can Reduce Sulphates to Sulphite
Target Organism Class Bacteria
Test Type Water Monitoring
For Use With (Application) Determine aerobic and anaerobic bacteria levels
Incubation Conditions 35°C
Incubation Time 5 Days
Manual/Automated Manual
Material Agar
No. per Pack 10/Pk.
Packaging Type Pack
Quantity 1 Ea.
Specimen Type Water, Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.