Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfate Standard, 1000 ppm SO4, Reagents

Catalog No. 01333939
Click to view available options
Quantity:
120 mL
500 mL

Catalog No. 01-333-939

Supplier: Reagents Holdings Llc CS014000500A

Only null left
Add to Cart
Add to Cart

Sulfate Standard, 1000 ppm SO4, Reagent, use for Sulfate analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfate Standard, 1000 ppm SO4
Packaging Poly Bottle
CAS 7732-18-5, 7757-82-6
Specific Gravity 1.0009
Physical Form Liquid
Color Colorless
pH 5.2 to 9.2
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Sulfate Analysis
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.