Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfate Standard, 1000 ppm SO4, Reagents

Catalog No. 01333940
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-940 120 mL
01-333-939 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-940 Supplier Reagents Holdings Llc Supplier No. CS014000120A
Only null left
Add to Cart
Add to Cart

Sulfate Standard, 1000 ppm SO4, Reagent, use for Sulfate analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfate Standard, 1000 ppm SO4
Packaging Poly Bottle
CAS 7732-18-5, 7757-82-6
Specific Gravity 1.0009
Physical Form Liquid
Color Colorless
pH 5.2 to 9.2
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Sulfate Analysis
Quantity 120 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.